Specifications
| Melting Point | 100.0°C to 102.0°C |
| Solubility | Soluble in methanol, chloroform and dimethyl sulfoxide |
| Quantity | 250 mg |
| Formula Weight | 222.67 |
| Chemical Name or Material | Acetamiprid |
Safety and Handling
Product Identifier
- Acetamiprid
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 2
- H301-Toxic if swallowed.
- H361d-Suspected of damaging the unborn child.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contain: Acetamiprid