missing translation for 'onlineSavingsMsg'
Learn More

ACCN1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-2531MFV450

 View more versions of this product

Product Code. 18718632

  • 570.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ACCN1 Polyclonal antibody specifically detects ACCN1 in Human, Rat samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ACCN1
Polyclonal
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Immunohistochemistry
mFluor Violet 450 SE
Acid-sensing ion channel 2, Amiloride-sensitive brain sodium channel, amiloride-sensitive cation channel 1, neuronal, ASIC2a, ASIC2Mammalian degenerin homolog, BNAC1, BNaC1ACCN, BNC1Amiloride-sensitive cation channel neuronal 1, degenerin, hBNaC1, MDEGBrain sodium channel 1, neuronal amiloride-sensitive cation channel 1
CSGPGRFMAREQ (amino acid residues 15-26).
0.1 mL
Apoptosis, Cell Biology, Neuroscience
40
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.