Learn More
Macherey-Nagel™ Accessories for Stainless Steel HPLC Columns
Click to view available options
Type:
Connector
Cutter Cone Fits
Locking Screw
Terminal Screw
Unit Size:
Each
Pack of 4
Pack of 5
Description
Stainless steel columns are most frequently used in HPLC. The material is corrosion resistant, pressure stable and easy to work mechanically.
Specifications
Specifications
| Type | Locking Screw |
| Color | White |
| For Use With (Application) | Locking Screw |
| Packaging Quantity | 4 Pack |
| For Use With | Analytical HPLC Columns |
| Material | Plastic |