missing translation for 'onlineSavingsMsg'
Learn More

ACBD6 Antibody (4A9D5E9), DyLight 680, Novus Biologicals™

Product Code. 18525930 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18525930 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18525930 Supplier Novus Biologicals Supplier No. NBP306614FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACBD6 Monoclonal antibody specifically detects ACBD6 in Human samples. It is validated for Western Blot, ELISA, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACBD6
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4A9D5E9
Conjugate DyLight 680
Dilution Western Blot, ELISA, Sandwich ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACBD6 (NP_115736.1; Met 1-Ala 282).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84320
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.