missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acadesine-15N4, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Acadesine-15N4 |
| Analyte or Component Names | Acadesine-15N4 |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H14 15N4 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside-15N4, AICAR-15N4, Acadesine-15N4, 5-Amino-1-β-D-ribofuranosyl-1H-imidazole-4-carboxamide-15N4, 5-Amino-4-imidazole carboxamide Riboside-15N4, Arasine-15N4, GP 1-110-15N4, NSC 105823-15N4 |
| InChI Formula | InChI=1S/C9H14N4O5/c10-7-4(8(11)17)12-2-13(7)9-6(16)5(15)3(1-14)18-9/h2-3,5-6,9,14-16H,1,10H2,(H2,11,17)/t3-,5-,6-,9-/m1/s1/i10+1,11+1,12+1,13+1 |
| Solution Type | Neat |
| SMILES | [15NH2]C(=O)c1[15n]c[15n]([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c1[15NH2] |
| IUPAC Name | 5-(^{15}N)azanyl-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl](1,3-^{15}N_{2})imidazole-4-(^{15}N)carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction