missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABH2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92472-0.02ml
This item is not returnable.
View return policy
Description
ABH2 Polyclonal antibody specifically detects ABH2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ABH2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 2OG-Fe(II) oxy DC1, ABH2FLJ99103, alkB, alkylation repair homolog 2 (E. coli), Alkylated DNA repair protein alkB homolog 2, alpha-ketoglutarate-dependent dioxygenase alkB homolog 2, EC 1.14.11.-, MGC90512, Oxy DC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-261 of human ALKBH2 (NP_001138847.1). MDRFLVKGAQGGLLRKQEEQEPTGEEPAVLGGDKESTRKRPRREAPGNGGHSAGPSWRHIRAEGLDCSYTVLFGKAEADEIFQELEKEVEYFTGALARVQVFGKWHSVPRKQATYGDAGLTYTFSGLTLSPKPWIPVLERIRDHVSGVTGQTFNFVLINRYKDGCDHIGEHRDDERELAPGSPIASVSFGACRDFVFRHKDSRGKSPSRRVAVVRLPLAHGSLLMMNHPTNTHWYHSLPVRKKVLAPRVNLTFRKILLTKK | |
| 0.02 mL | |
| Direct Reversal of DNA Damage | |
| 121642 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction