missing translation for 'onlineSavingsMsg'
Learn More

ABCG2, Mouse anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16120616
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16120616 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16120616 Supplier Abnova Supplier No. H00009429A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Sequence: MSSSNVEVFIPVSQGNTNGFPATVSNDLKAFTEGAVLSFHNICYRVKLKSGFLPCRKPVEKEILSNINGIMKPGLNAILGPTGGGKSSLLDVLAARKDPSGLSGDVLING

Specifications

Antigen ABCG2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 50 % glycerol
Gene ATP-binding cassette, sub-family G (WHITE), member 2
Gene Accession No. NM_004827
Gene Alias ABC15/ABCP/BCRP/BCRP1/BMDP/CD338/CDw338/EST157481/MGC102821/MRX/MXR/MXR1
Gene Symbols ABCG2
Host Species Mouse
Immunogen ABCG2 (NP_004818, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9429
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.