missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ABCB11 Antibody (BSEP/7534) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27037AF700
This item is not returnable.
View return policy
Description
ABCB11 Monoclonal antibody specifically detects ABCB11 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| ABCB11 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| ABC member 16, MDR/TAP subfamily, ABC16, ATP-binding cassette sub-family B member 11, ATP-binding cassette, sub-family B (MDR/TAP), member 11, bile salt export pump, BSEP, BSEPPFIC2, EC 3.6.3, EC 3.6.3.44, PFIC-2, PGY4, progressive familial intrahepatic cholestasis 2, sister p-glycoprotein, SPGPBRIC2 | |
| Recombinant fragment (around aa600-800) of human ABCB11 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 8647 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| BSEP/7534 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction