missing translation for 'onlineSavingsMsg'
Learn More

Abcam Ro 3306, cyclin-dependent kinase (Cdk1) inhibitor, 1 mg

Product Code. 32109128
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109128 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109128 Supplier Abcam Supplier No. ab1414911mg
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody, with 6 citations.

  • Storage Buffer: Constituents: 100% (5Z)-5-(quinolin-6-ylmethylidene)-2-(thiophen-2-ylmethylimino)-1,3-thiazolidin-4-one
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen CID 11631681
Conjugate Unconjugated
Formulation Constituents: 100% (5Z)-5-(quinolin-6-ylmethylidene)-2-(thiophen-2-ylmethylimino)-1,3-thiazolidin-4-one
Gene CID 11631681
Gene Symbols CID 11631681
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Target Species Non-species specific
Product Type Antibody (CID 11631681)
Format Vial
Shipping Condition Ambient
Target Gene CID 11631681
Target Entry CID 11631681
Target Molecule CID 11631681
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.