missing translation for 'onlineSavingsMsg'
Learn More

Abcam Recombinant Human Prostate Specific Antigen protein, 50 ug

Product Code. 32148311
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32148311 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32148311 Supplier Abcam Supplier No. ab28343050ug
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Recombinant protein, suitable for SDS-PAGE, Mass Spectrometry, HPLC.

  • Species: Human
  • Gene Symbol: KLK3
  • Applications: SDS-PAGE, Mass Spectrometry, HPLC
  • Form: Lyophilized
  • Purity: ≥95%
  • Buffer: pH: 7.4, Constituents: 10.26% Trehalose, 0.727% Dibasic monohydrogen potassium phosphate, 0.248% Potassium phosphate monobasic

Specifications

Accession Number P07288
For Use With (Application) SDS-PAGE, Mass Spectrometry, HPLC
Formulation pH: 7.4, Constituents: 10.26% Trehalose, 0.727% Dibasic monohydrogen potassium phosphate, 0.248% Potassium phosphate monobasic
Gene ID (Entrez) 354
Name Recombinant Human Prostate Specific Antigen protein
Quantity 50 μg
Regulatory Status RUO
Gene Alias APS; KLK3; Prostate-specific antigen; PSA; Gamma-seminoprotein; Kallikrein-3; P-30 antigen; Semenogelase; Seminin
Common Name KLK3
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute in PBS
Buffer pH: 7.4, Constituents: 10.26% Trehalose, 0.727% Dibasic monohydrogen potassium phosphate, 0.248% Potassium phosphate monobasic
Form Lyophilized
Purity or Quality Grade ≥95%
Synonym APS; KLK3; Prostate-specific antigen; PSA; Gamma-seminoprotein; Kallikrein-3; P-30 antigen; Semenogelase; Seminin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.