missing translation for 'onlineSavingsMsg'
Learn More

Abcam Recombinant human BMP4 protein (Active), 20 ug

Product Code. 32153055
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32153055 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32153055 Supplier Abcam Supplier No. ab23829820ug
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Recombinant protein, suitable for SDS-PAGE, Functional Study, with 2 citations.

  • Species: Human
  • Gene Symbol: BMP4
  • Applications: SDS-PAGE, Functional Study
  • Form: Lyophilized
  • Purity: >95%
  • Buffer: Constituents: 0.1% Trifluoroacetic acid

Specifications

Accession Number P12644
For Use With (Application) SDS-PAGE, Functional Study
Formulation Constituents: 0.1% Trifluoroacetic acid
Gene ID (Entrez) 652
Name Recombinant human BMP4 protein (Active)
Quantity 20 μg
Storage Requirements Avoid freeze / thaw cycle
Regulatory Status RUO
Gene Alias BMP2B; DVR4; BMP4; Bone morphogenetic protein 4; BMP-4; Bone morphogenetic protein 2B; BMP-2B
Common Name BMP4
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 0.1 mg/mL in water
Buffer Constituents: 0.1% Trifluoroacetic acid
Content And Storage Avoid freeze / thaw cycle
Form Lyophilized
Purity or Quality Grade >95%
Synonym BMP2B; DVR4; BMP4; Bone morphogenetic protein 4; BMP-4; Bone morphogenetic protein 2B; BMP-2B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.