missing translation for 'onlineSavingsMsg'
Learn More

Abcam Recombinant human ANGPTL3 protein, 50 ug

Product Code. 32150098
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32150098 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32150098 Supplier Abcam Supplier No. ab17602850ug
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Recombinant protein, suitable for ELISA, HPLC, SDS-PAGE, Functional Study.

  • Species: Human
  • Gene Symbol: ANGPTL3
  • Applications: ELISA, HPLC, SDS-PAGE, Functional Study
  • Form: Lyophilized
  • Purity: >98%

Specifications

Accession Number Q9Y5C1
For Use With (Application) ELISA, HPLC, SDS-PAGE, Functional Study
Gene ID (Entrez) 27329
Name Recombinant human ANGPTL3 protein
Quantity 50 μg
Storage Requirements Avoid freeze / thaw cycle
Regulatory Status RUO
Gene Alias ANGPT5; UNQ153/PRO179; ANGPTL3; Angiopoietin-related protein 3; Angiopoietin-5; Angiopoietin-like protein 3; ANG-5
Common Name ANGPTL3
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage Avoid freeze / thaw cycle
Form Lyophilized
Purity or Quality Grade >98%
Synonym ANGPT5; UNQ153/PRO179; ANGPTL3; Angiopoietin-related protein 3; Angiopoietin-5; Angiopoietin-like protein 3; ANG-5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.