Learn More
Abcam Rat monoclonal [SB74g] Anti-Mouse IgG2b gamma chain (Alexa Fluor™ 488), 250 ug
Product Code. 32109989 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109989 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32109989 Supplier Abcam Supplier No. ab172326250ug
 Restricted Item
Estimated Shipment: 03-12-2026
Rat Monoclonal antibody, suitable for Flow Cyt, ELISA, IHC-P, ICC/IF, with 2 citations.
  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Applications: Flow Cyt, ELISA, IHC-P, ICC/IF
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
  • Form: Liquid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SB74g
Conjugate Unconjugated
Gene Mouse IgG2b
Host Species Rat
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Mouse IgG2b
Validated Application abrv Flow Cyt, ELISA, IHC-P, ICC/IF