missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rabbit F(ab')2 Anti-Dog IgG H&L (Alkaline Phosphatase), 500 uL

Product Code. 32119786 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119786 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32119786 Supplier Abcam Supplier No. ab136764500uL
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal secondary antibody, suitable for FuncS, ELISA, IHC-P, WB.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: FuncS, ELISA, IHC-P, WB, IHC-Fr, ICC
  • Species Reactivity: Dog
  • Storage Buffer: Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 48.9% Tris buffered saline, 1% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 48.9% Tris buffered saline, 1% BSA
Host Species Rabbit
Purification Method Affinity purification Immunogen
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Dog.
Target Species Dog
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Dog
Validated Application abrv FuncS, ELISA, IHC-P, WB, IHC-Fr, ICC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.