missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rabbit Anti-Mouse IgG H&L (HRP), 1 mg

Product Code. 32115056 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115056 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32115056 Supplier Abcam Supplier No. ab67281mg
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal secondary antibody, suitable for ELISA, IHC-Fr, IHC-P, WB, with over 1,000 citations.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ELISA, IHC-Fr, IHC-P, WB
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.01% Gentamicin sulphate, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
  • Form: Liquid
  • Purification: Affinity purification

Specifications

Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Preservative: 0.01% Gentamicin sulphate, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Gene Symbols Ighg1
Host Species Rabbit
Purification Method Affinity purification
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv ELISA, IHC-Fr, IHC-P, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.