missing translation for 'onlineSavingsMsg'
Learn More

Abcam Mouse monoclonal [7C2] Anti-Chicken IgY H&L, 250 uL

Product Code. 32110653 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
250µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110653 250 μL 250µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32110653 Supplier Abcam Supplier No. ab8922250uL
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal antibody, suitable for WB, IP, with 1 citations.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: WB, IP
  • Species Reactivity: Chicken
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: 0.1% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein G

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7C2
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: 0.1% BSA
Host Species Mouse
Purification Method Affinity purification Protein G
Quantity 250 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Chicken.
Target Species Chicken
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Ck
Target Molecule Chicken IgY
Validated Application abrv WB, IP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.