missing translation for 'onlineSavingsMsg'
Learn More

Abcam Mouse monoclonal [4E3] Anti-Human IgG1 hinge heavy chain, 250 ug

Product Code. 32110408 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110408 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32110408 Supplier Abcam Supplier No. ab99771250ug
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal antibody, suitable for ICC/IF, Flow Cyt, ELISA.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: ICC/IF, Flow Cyt, ELISA
  • Species Reactivity: Human
  • Storage Buffer: pH: 8.2, Constituents: 100% Borate buffered saline
  • Form: Liquid
  • Purification: Affinity purification Protein G

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 4000
Conjugate Unconjugated
Formulation pH: 8.2, Constituents: 100% Borate buffered saline
Gene IGHG1
Gene Symbols IGHG1
Host Species Mouse
Purification Method Affinity purification Protein G
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Target Molecule IGHG1, Human
Validated Application abrv ICC/IF, Flow Cyt, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.