missing translation for 'onlineSavingsMsg'
Learn More

Abcam Goat F(ab')2 Anti-Rat IgG - (Fab')2 (HRP), pre-adsorbed, 500 uL

Product Code. 32110147 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110147 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32110147 Supplier Abcam Supplier No. ab6517500uL
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal secondary antibody, suitable for WB, Dot, EM, IHC, with 3 citations.

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: WB, Dot, EM, IHC, ICC, ELISA
  • Species Reactivity: Rat
  • Storage Buffer: Constituents: 1.5% BSA, 1.46% Sodium chloride, 0.164% Sodium phosphate
  • Form: Liquid

Specifications

Applications Western Blot, Dot Blot, Electron Microscopy, Immunohistochemistry, Immunocytochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation Constituents: 1.5% BSA, 1.46% Sodium chloride, 0.164% Sodium phosphate
Host Species Goat
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Reactivity abrv Rt
Validated Application abrv WB, Dot, EM, IHC, ICC, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.