missing translation for 'onlineSavingsMsg'
Learn More

Abcam Donkey Anti-Sheep IgG H&L, 1 mg

Product Code. 32119837 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119837 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32119837 Supplier Abcam Supplier No. ab68951mg
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal secondary antibody, suitable for Conjugation, IHC-Fr, IHC-P, IP, with 2 citations.

  • Host Species: Donkey
  • Clonality: Polyclonal
  • Applications: Conjugation, IHC-Fr, IHC-P, IP, ID, WB, ELISA, ICC/IF
  • Species Reactivity: Sheep
  • Storage Buffer: pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 0.88% Sodium chloride, 0.424% Potassium phosphate solution
  • Form: Liquid
  • Purification: Affinity purification

Specifications

Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Host Species Donkey
Purification Method Affinity purification
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Sheep.
Target Species Sheep
Product Type Polyclonal Secondary Antibody
Form Liquid
Shipping Condition Blue Ice
Reactivity abrv Sh
Validated Application abrv Conjugation, IHC-Fr, IHC-P, IP, ID, WB, ELISA, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.