missing translation for 'onlineSavingsMsg'
Learn More

Abcam Donkey Anti-Mouse IgG H&L (10nm Gold), 1250 uL

Product Code. 32117500 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1250 μL
Unit Size:
1250µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32117500 1250 μL 1250µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32117500 Supplier Abcam Supplier No. ab395931250uL
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal secondary antibody, suitable for EM, with 9 citations.

  • Host Species: Donkey
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: EM
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.097% Sodium azide, Constituents: PBS, 1% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Electron Microscopy
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.097% Sodium azide, Constituents: PBS, 1% BSA
Gene Symbols Ighg1
Host Species Donkey
Purification Method Affinity purification Immunogen
Quantity 1250 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv EM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.