missing translation for 'onlineSavingsMsg'
Learn More

Abcam Donkey Anti-Goat IgG H&L (Cy5.5 ™) preadsorbed, 500 ug

Product Code. 32108911 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32108911 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32108911 Supplier Abcam Supplier No. ab6951500ug
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal secondary antibody, suitable for Flow Cyt, ICC, IHC-Fr, IHC-P, with 11 citations.

  • Host Species: Donkey
  • Clonality: Polyclonal
  • Applications: Flow Cyt, ICC, IHC-Fr, IHC-P, ELISA, ICC/IF
  • Species Reactivity: Goat
  • Storage Buffer: pH: 6.5, Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.87% Sodium chloride, 0.42% Tripotassium orthophosphate
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 6.5, Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.87% Sodium chloride, 0.42% Tripotassium orthophosphate
Host Species Donkey
Purification Method Affinity purification Immunogen
Quantity 500 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Goat.
Target Species Goat
Product Type Polyclonal Secondary Antibody
Form Liquid
Shipping Condition Blue Ice
Reactivity abrv Gt
Validated Application abrv Flow Cyt, ICC, IHC-Fr, IHC-P, ELISA, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.