missing translation for 'onlineSavingsMsg'
Learn More

Abcam Cell Cycle (pCdk/pHH3/Actin) WB Cocktail, 200 uL

Product Code. 32119910 Shop All Abcam Products
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119910 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32119910 Supplier Abcam Supplier No. ab136810200uL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for WB, with 17 citations.

  • Applications: WB
  • Species Reactivity: Mouse and Human
  • Storage Buffer: Preservative: 0.01% Sodium azide, Constituents: 40% Glycerol (glycerin, glycerine), 0.7% Sodium chloride, 0.61% Tris, 0.05% BSA
  • Form: Liquid

Specifications

Applications Western Blot
Conjugate Unconjugated
Formulation Preservative: 0.01% Sodium azide, Constituents: 40% Glycerol (glycerin, glycerine), 0.7% Sodium chloride, 0.61% Tris, 0.05% BSA
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Mouse and Human.
Target Species Mouse, Human
Product Type Secondary Antibody
Form Liquid
Shipping Condition Blue Ice
Reactivity abrv Ms, Hu
Validated Application abrv WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.