missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-YTHDC1 antibody, 100 uL

Product Code. 31936886
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31936886 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31936886 Supplier Abcam Supplier No. ab122340100uL
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal antibody, suitable for IHC-P, WB, ICC/IF, with 36 citations.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: IHC-P, WB, ICC/IF
  • Species Reactivity: Human
  • Storage Buffer: pH: 7.2, Preservative: 0.02% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine)
  • Form: Liquid
  • Purification: Affinity purification Immunogen

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen YTHDC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.2, Preservative: 0.02% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine)
Gene YTHDC1
Gene Symbols YTHDC1
Host Species Rabbit
Purification Method Affinity purification Immunogen
Quantity 100 μL
Regulatory Status RUO
Chains or Fragments IgG
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Polyclonal Antibody (YTHDC1)
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Hu
Target Gene YTHDC1
Target Entry YTHDC1
Target Molecule YTHDC1, Human
Technique IHC-P, WB, ICC/IF
Validated Application abrv IHC-P, WB, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.