missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-SOCS3 antibody [EPR24090-74], 20 uL

Product Code. 32093443 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
Unit Size:
20µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32093443 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32093443 Supplier Abcam Supplier No. ab28088420uL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal secondary antibody, suitable for ICC/IF, IP, Flow Cyt, WB, with 21 citations.

  • Host Species: Rabbit
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: ICC/IF, IP, Flow Cyt, WB, IHC-P
  • Species Reactivity: Mouse, Rat and Human
  • Storage Buffer: Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein A

Specifications

Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EPR24090-74
Conjugate Unconjugated
Formulation Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene Symbols SOCS3
Host Species Rabbit
Purification Method Affinity purification Protein A
Quantity 20 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse, Rat and Human.
Target Species Mouse, Rat, Human
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt, Hu
Validated Application abrv ICC/IF, IP, Flow Cyt, WB, IHC-P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.