missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-Perilipin-2 antibody [EPR28638-122], 20 uL

Product Code. 32072888
Change view
Click to view available options
Quantity:
20 μL
Unit Size:
20µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32072888 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32072888 Supplier Abcam Supplier No. ab32464320uL
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Monoclonal antibody, suitable for Dot, ICC/IF, IHC-P.

  • Host Species: Rabbit
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: Dot, ICC/IF, IHC-P
  • Species Reactivity: Human and Mouse
  • Storage Buffer: pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen PLIN2
Applications Dot Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EPR28638-122
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene PLIN2
Gene Symbols PLIN2
Host Species Rabbit
Purification Method Affinity purification Protein A
Quantity 20 μL
Regulatory Status RUO
Chains or Fragments IgG
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Human and Mouse.
Target Species Human, Mouse
Product Type Monoclonal Antibody (PLIN2)
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Hu, Ms
Target Gene PLIN2
Target Entry PLIN2
Target Molecule PLIN2, Human, Mouse
Technique Dot, ICC/IF, IHC-P
Validated Application abrv Dot, ICC/IF, IHC-P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.