missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-Oct4 antibody, 100 ug

Product Code. 32047240
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32047240 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32047240 Supplier Abcam Supplier No. ab19857100ug
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal antibody, suitable for IP, WB, ICC/IF, with over 600 citations.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: IP, WB, ICC/IF
  • Species Reactivity: Mouse and Human
  • Storage Buffer: pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 1% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen POU5F1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 1% BSA
Gene POU5F1
Gene Symbols POU5F1
Host Species Rabbit
Purification Method Affinity purification Protein A
Quantity 100 μg
Regulatory Status RUO
Chains or Fragments IgG
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse and Human.
Target Species Mouse, Human
Product Type Polyclonal Antibody (POU5F1)
Form Liquid
Format Vial
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms, Hu
Target Gene POU5F1
Target Entry POU5F1
Target Molecule POU5F1, Mouse, Human
Technique IP, WB, ICC/IF
Validated Application abrv IP, WB, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.