missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-mCherry antibody [1C51], 100 uL

Product Code. 32056075 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32056075 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32056075 Supplier Abcam Supplier No. ab125096100uL
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal antibody, suitable for WB, ICC/IF, IHC-P, with over 200 citations.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG2a
  • Applications: WB, ICC/IF, IHC-P
  • Storage Buffer: Preservative: 0.03% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
  • Form: Liquid
  • Purification: Affinity purification Protein G

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen mCherry
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1C51
Conjugate Unconjugated
Formulation Preservative: 0.03% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene mCherry
Gene Symbols mCherry
Host Species Mouse
Purification Method Affinity purification Protein G
Quantity 100 μL
Regulatory Status RUO
Chains or Fragments IgG2a
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Target Species Non-species specific
Product Type Monoclonal Antibody (mCherry)
Form Liquid
Format Vial
Isotype IgG2a
Host abrv Ms
Shipping Condition Blue Ice
Target Gene mCherry
Target Entry mCherry
Target Molecule mCherry
Technique WB, ICC/IF, IHC-P
Validated Application abrv WB, ICC/IF, IHC-P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.