missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-MAP2 antibody - Neuronal Marker, 25 uL

Product Code. 32047536 Shop All Abcam Products
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
25µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32047536 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32047536 Supplier Abcam Supplier No. ab539225uL
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken Polyclonal antibody, suitable for IHC-P, ICC/IF, WB, with over 1,000 citations.

  • Host Species: Chicken
  • Clonality: Polyclonal
  • Isotype: IgY
  • Applications: IHC-P, ICC/IF, WB
  • Species Reactivity: Mouse and Rat
  • Storage Buffer: Preservative: 0.032% Sodium azide, Constituents: PBS
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen MAP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.032% Sodium azide, Constituents: PBS
Gene MAP2
Gene Symbols MAP2
Host Species Chicken
Quantity 25 μL
Regulatory Status RUO
Chains or Fragments IgY
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse and Rat.
Target Species Mouse, Rat
Product Type Polyclonal Antibody (MAP2)
Form Liquid
Format Vial
Isotype IgY
Host abrv Ch
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt
Target Gene MAP2
Target Entry MAP2
Target Molecule MAP2, Mouse, Rat
Technique IHC-P, ICC/IF, WB
Validated Application abrv IHC-P, ICC/IF, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.