missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-Lrp2 / Megalin antibody, 100 uL

Product Code. 32046035 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32046035 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32046035 Supplier Abcam Supplier No. ab236244100uL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal secondary antibody, suitable for IHC-P, ICC/IF, with 3 citations.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: IHC-P, ICC/IF
  • Species Reactivity: Human
  • Storage Buffer: pH: 7.4, Preservative: 0.03% Proclin 300, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
  • Form: Liquid
  • Purification: Affinity purification Protein G

Specifications

Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.03% Proclin 300, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene Symbols LRP2
Host Species Rabbit
Purification Method Affinity purification Protein G
Quantity 100 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv IHC-P, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.