missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-IRS1 (phospho S307) antibody, 100 ug

Product Code. 32056984 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32056984 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32056984 Supplier Abcam Supplier No. ab5599100ug
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal antibody, suitable for ICC, WB, with 18 citations.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ICC, WB
  • Species Reactivity: Mouse and Human
  • Storage Buffer: Preservative: 0.05% Sodium azide, Constituents: PBS, 0.1% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen IRS1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.05% Sodium azide, Constituents: PBS, 0.1% BSA
Gene IRS1
Gene Symbols IRS1
Host Species Rabbit
Purification Method Affinity purification Immunogen
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse and Human.
Target Species Mouse, Human
Product Type Polyclonal Antibody (IRS1)
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms, Hu
Target Modification State phospho S307
Target Gene IRS1
Target Entry IRS1
Target Molecule IRS1, Mouse, Human
Technique ICC, WB
Validated Application abrv ICC, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.