missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-Insulin antibody [K36aC10], 100 ug

Product Code. 32097191 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32097191 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32097191 Supplier Abcam Supplier No. ab6995100ug
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal secondary antibody, suitable for IHC-P, with over 100 citations.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: IHC-P
  • Species Reactivity: Human
  • Storage Buffer: Preservative: 0.05% Sodium azide, Constituents: PBS, 0.05% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein A/G

Specifications

Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone K36aC10
Conjugate Unconjugated
Formulation Preservative: 0.05% Sodium azide, Constituents: PBS, 0.05% BSA
Gene Symbols INS
Host Species Mouse
Purification Method Affinity purification Protein A/G
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG1
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv IHC-P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.