Learn More
Abcam Anti-Human IgG4 pFc' [HP6023] (PE), 100 ug
Artikelnummer. 32119915 Shop alle Abcam produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
100 μg
Pakningsstørrelse:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
32119915 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 32119915 Leverandør Abcam Leverandørnr. ab99819100ug
 Restricted Item
Estimated Shipment: 03-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Anti-Human IgG4 pFc' [HP6023] (PE)
  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG3
  • Applications: Dot, ELISA, ICC/IF, IHC-Fr, WB
  • Species Reactivity: Human
  • Storage Buffer: PBS, Sucrose
  • Form: Liquid
  • Purification: Affinity purification Protein A
Tekniske data
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone HP6023
Conjugate Unconjugated
Formulation pH: 7.3, Preservative: 0.093% Sodium azide, Constituents: PBS, 1% Sucrose
Gene IGHG4
Gene Symbols IGHG4
Host Species Mouse
Purification Method Affinity purification Protein A
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG3
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Target Molecule IGHG4, Human
Validated Application abrv Dot, ELISA, WB, IHC-Fr, ICC/IF
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.