Learn More
Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates
GREENER_CHOICE
Product Code. 10276963 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue, White Bottom
Green, Clear
Red, Clear
White
Yellow, Clear
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10276963 Blue, White Bottom Pack of 25
10308713 White Pack of 25
10464393 Green, Clear Pack of 25
10777294 Red, Clear Pack of 25
10552065 Yellow, Clear Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10276963 Supplier Eppendorf™ Supplier No. 0030132530
In Stock
Add to basket
Add to basket
Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors. Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates feature a one-piece design-combining polycarbonate and polypropylene for optimum performance.
  • Improved well-to-well tolerance
  • Semi-skirt for labelling
  • Extremely thin-walled for optimum heat transfer into the sample
  • Cut-away corner and alphanumeric grid-referencing
  • Raised well rims for effective sealing and reduced cross-contamination risk
  • 250μL well volume (when used with cap strips)
TRUSTED_SUSTAINABILITY
Specifications
Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color Blue, White Bottom
Product Type 96-well PCR Plate
No. of Wells 96
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Profile 96 Wells, Semi-Skirted
Quantity 25/Pk.
Temperature Range (Metric) 121°C