Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | White, Clear Bottom |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Sterile |
| Well Shape | Conical Bottom (V-well) |
| Quantity | 240/Pk. |
| Skirt Color | White |
| Well Color | Clear |