Learn More
Tocris Bioscience™ 9-Phenanthrol
Product Code. 18776667
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18776667 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18776667 Supplier Tocris Bioscience™ Supplier No. 4999/50
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Selective TRPM4 blocker
9-Phenanthrol is a selective TRPM4 blocker (IC50 = 20 μM in HEK293 cells). Exhibits no effect on CFTR or TRPM5 (at 0.25 and 1 mM respectively). Abolishes arrhythmias induced by hypoxia in a mouse heart model.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 mg
Product Type 9-Phenanthrol
Molecular Weight (g/mol) 194.23
Inhibitors 9-Phenanthrol
For Research Use Only
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.