missing translation for 'onlineSavingsMsg'
Learn More

9-Oxofolic Acid, TRC

Product Code. 30007455
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007455 50 mg 50mg
30011254 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007455 Supplier Toronto Research Chemicals Supplier No. TRCO85685050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39707-61-4
Molecular Formula C19H17N7O7
Molecular Weight (g/mol) 455.38
Synonym N-[4-[[(2-Amino-1,4-dihydro-4-oxo-6-pteridinyl)carbonyl]amino]benzoyl]-L-glutamic Acid
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridine-6-carbonyl)amino]benzoyl]amino]pentanedioic acid
SMILES O=C1C2=NC(C(NC3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=O)=CNC2=NC(N)=N1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H17N7O7/c20-19-25-14-13(17(31)26-19)23-11(7-21-14)16(30)22-9-3-1-8(2-4-9)15(29)24-10(18(32)33)5-6-12(27)28/h1-4,7,10H,5-6H2,(H,22,30)(H,24,29)(H,27,28)(H,32,33)(H3,20,21,25,26,31)/t10-/m0/s1
Formula Weight 455.1189
Percent Purity >95%
Chemical Name or Material 9-Oxofolic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.