CAS RN 39707-61-4
CAS RN 39707-61-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-Oxofolic Acid, TRC
CAS: 39707-61-4 Molecular Formula: C19H17N7O7 Molecular Weight (g/mol): 455.38 Synonym: N-[4-[[(2-Amino-1,4-dihydro-4-oxo-6-pteridinyl)carbonyl]amino]benzoyl]-L-glutamic Acid IUPAC Name: (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridine-6-carbonyl)amino]benzoyl]amino]pentanedioic acid SMILES: O=C1C2=NC(C(NC3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=O)=CNC2=NC(N)=N1