missing translation for 'onlineSavingsMsg'
Learn More

8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Product Code. 15429015
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429015 250 mg 250mg
15419015 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429015 Supplier Thermo Scientific Alfa Aesar Supplier No. H35387.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1018828-72-2
Molecular Formula C9H5F3N2O2
Molecular Weight (g/mol) 230.15
MDL Number MFCD09995161
InChI Key QRXSLIVFFDRFEB-UHFFFAOYSA-N
Synonym 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo-1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylicacid, 8-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylic acid
PubChem CID 59443885
IUPAC Name 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F

Specifications

Melting Point ∽242°C (decomposition)
Quantity 250 mg
Formula Weight 230.15 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.