CAS RN 1018828-72-2
CAS RN 1018828-72-2
IUPAC Name:
8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Synonyms:
8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Molecular Weight (g/mol):
230.15
Molecular Formula:
C9H5F3N2O2
InChi Key:
QRXSLIVFFDRFEB-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F
1
–
1
of
1
results
1
8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™
CAS: 1018828-72-2 Molecular Formula: C9H5F3N2O2 Molecular Weight (g/mol): 230.15 MDL Number: MFCD09995161 InChI Key: QRXSLIVFFDRFEB-UHFFFAOYSA-N Synonym: 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid,8-trifluoromethyl imidazo-1,2-a pyridine-2-carboxylic acid,8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylicacid,8-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylic acid PubChem CID: 59443885 IUPAC Name: 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid SMILES: OC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F