Learn More
Applied Biosystems™ 7900HT TaqMan™ Low Density Array Instrument Verification RNase P Kit
Shop alle Applied Biosystems produkterBeskrivelse
The 7900HT TaqMan™ Low Density Array Instrument Verification RNase P kit contains an empty TaqMan™ Low Density Array and 8 vials of RNase P detection reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each vial contains premixed reagents (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template. The 7900HT TaqMan™ Low Density Array Instrument Verification RNase P kit must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems™ recommends that the performance of your instrument be verified every six months.
Tekniske data
Tekniske data
| Content And Storage | Contains an empty TaqMan™ Low Density Array and 8 vials of RNase P detection reagents. |
| For Use With (Equipment) | 7900HT System |
| Product Type | Spectral Calibration Kit |
| For Use With (Application) | Real time PCR (qPCR) |
| Description | Fluorescent Detection |
| Quantity | 1 Kit |
| Calibration Type | Instrument Verification Plate, RNase P Calibration |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.