Learn More
700 mOsm/kg Osmolality Standard, Reagecon
BIOPHARMA
Product Code. 16368185 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
12 x 5 mL
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16368185 12 x 5 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16368185 Supplier Reagecon Supplier No. REOSM700
  • ONLINE EXCLUSIVE PRICE
    142.00€
    140.32€ / Pack of 12
    Save 1.68€
    1% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Edge
Add to basket
Used to calibrate, control or validate osmometers
  • Manufactured in compliance with European, United States and Chinese Pharmacopoeia guidelines
  • Tested by ISO 17025 accredited test method
  • ISO 17025 accreditation for calibration of laboratory balances
  • Prepared gravimetrically on weight/weight basis
Specifications
Packaging Amber glass ampoule
Physical Form Liquid
Technique Calibration, Control or Validate Osmometer
For Use With (Equipment) Osmometer
Type Osmolality Standard
Quantity 12 x 5 mL