missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole, 99%, Thermo Scientific™

Product Code. 11420080
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420080 1 g 1g
11410080 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420080 Supplier Thermo Scientific Alfa Aesar Supplier No. H50592.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164736-47-4
Molecular Formula C11H10BrN
Molecular Weight (g/mol) 236.112
MDL Number MFCD00239009
InChI Key XKDWSCYYHKJAGK-UHFFFAOYSA-N
Synonym 7-bromo-1,2,3,4-tetrahydrocyclopenta b indole, 7-bromo-1,2,3,4-tetrahydrocyclopent b indole, 7-bromo-1h,2h,3h,4h-cyclopenta b indole, cyclopent b indole, 7-bromo-1,2,3,4-tetrahydro, 7-bromo-1,2,3-trihydrocyclopenta 1,2-b indole, bio6e6, acmc-20a4tv
PubChem CID 700805
IUPAC Name 7-bromo-1,2,3,4-tetrahydrocyclopenta[b]indole
SMILES C1CC2=C(C1)NC3=C2C=C(C=C3)Br

Specifications

Quantity 1 g
Formula Weight 236.11
Percent Purity 99%
Chemical Name or Material 7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.