accessibility menu, dialog, popup

UserName

HNBr

CAS RN 164736-47-4

IUPAC Name: 7-bromo-1H,2H,3H,4H-cyclopenta[b]indole
Synonyms: 7-bromo-1H,2H,3H,4H-cyclopenta[b]indole
Molecular Weight (g/mol): 236.11
Molecular Formula: C11H10BrN
InChi Key: XKDWSCYYHKJAGK-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC3=C(CCC3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results