missing translation for 'onlineSavingsMsg'
Learn More

7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride (Ethylenediamine Compound Hydrochloride), Mikromol™

Product Code. 17932406
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932406 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932406 Supplier Mikromol Supplier No. MM0018.120025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride (Ethylenediamine Compound Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desethylene Ciprofloxacin Hydrochloride
CAS 528851-31-2
Analyte or Component Names Desethylene Ciprofloxacin Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C15 H16 F N3 O3 . Cl H
Certifications/Compliance ISO 9001
Synonym 7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid hydrochloride (1:1), 7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid monohydrochloride, 7-(2-Aminoethylamino)-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride, Ciprofloxacin Ethylenediamine Compound Hydrochloride, Ciprofloxacin Ethylenediamine Analog, Desethylene Ciprofloxacin Hydrochloride, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-[(2-aminoethyl)-amino]-3-quinolinecarboxylic acid hydrochloride, Ciprofloxacin Hydrochloride Impurity C as Hydrochloride, Ciprofloxacin Impurity C as Hydrochloride
InChI Formula InChI=1S/C15H16FN3O3.ClH/c16-11-5-9-13(6-12(11)18-4-3-17)19(8-1-2-8)7-10(14(9)20)15(21)22;/h5-8,18H,1-4,17H2,(H,21,22);1H
SMILES Cl.NCCNc1cc2N(C=C(C(=O)O)C(=O)c2cc1F)C3CC3
IUPAC Name 7-(2-aminoethylamino)-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 341.77
Quantity 25 mg
Formula Weight 341.0942 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.