missing translation for 'onlineSavingsMsg'
Learn More

6Alpha-9-Difluoroprednisolone-17-Carboxylate, TRC

Product Code. 30385372
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385372 50 mg
30371901 100 mg
30370119 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385372 Supplier Toronto Research Chemicals Supplier No. TRCD44714550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1234557-20-0
Molecular Formula C20H24F2O5
Molecular Weight (g/mol) 382.4
Synonym 6A-9-Difluoroprednisolone-17-Carboxylate
SMILES OC(=O)c1c(C)n(nc1)c2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H24F2O5/c1-17-5-3-10(23)7-13(17)14(21)8-12-11-4-6-19(27,16(25)26)18(11,2)9-15(24)20(12,17)22/h3,5,7,11-12,14-15,24,27H,4,6,8-9H2,1-2H3,(H,25,26)
Chemical Name or Material 6alpha-9-Difluoroprednisolone-17-Carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.