CAS RN 1234557-20-0
CAS RN 1234557-20-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6Alpha-9-Difluoroprednisolone-17-Carboxylate, TRC
CAS: 1234557-20-0 Molecular Formula: C20H24F2O5 Molecular Weight (g/mol): 382.4 Synonym: 6A-9-Difluoroprednisolone-17-Carboxylate SMILES: OC(=O)c1c(C)n(nc1)c2ccccc2