missing translation for 'onlineSavingsMsg'
Learn More

6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-diene-17beta-carbothioic Acid, Mikromol™

Product Code. 30052054
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30052054

Brand: Mikromol MM1244.140025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-diene-17beta-carbothioic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6a,9a-Difluoro-11beta-hydroxy-16a-methyl-3-oxo-17a-(propionyloxy)-androsta-1,4-diene-17beta-carbothioic Acid
CAS 80474-45-9
Analyte or Component Names 6a,9a-Difluoro-11beta-hydroxy-16a-methyl-3-oxo-17a-(propionyloxy)-androsta-1,4-diene-17beta-carbothioic Acid
Shipping Condition Room Temperature
Molecular Formula C24 H30 F2 O5 S
Certifications/Compliance ISO 9001
Synonym Androsta-1,4-diene-17-carbothioic acid, 6,9-difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)-, (6alpha,11beta,16alpha,17alpha)-, (6alpha,11beta,16alpha,17alpha)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid, 6a,9a-Difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17alpha-propionyloxyandrosta-1,4-diene-17beta-carbothioic acid, CCI 22363, Fluticasone related
InChI Formula InChI=1S/C24H30F2O5S/c1-5-19(29)31-24(20(30)32)12(2)8-14-15-10-17(25)16-9-13(27)6-7-21(16,3)23(15,26)18(28)11-22(14,24)4/h6-7,9,12,14-15,17-18,28H,5,8,10-11H2,1-4H3,(H,30,32)/t12-,14+,15+,17+,18+,21+,22+,23+,24+/m1/s1
SMILES CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)S
IUPAC Name (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carbothioic S-acid
Molecular Weight (g/mol) 468.55
Quantity 25 mg
Formula Weight 468.1782 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.