missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-17-[(methylsulfanyl)carbonyl]-3-oxoandrosta-1,4-dien-17alpha-yl Propanoate, Mikromol™
Description
6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-17-[(methylsulfanyl)carbonyl]-3-oxoandrosta-1,4-dien-17alpha-yl Propanoate
Specifications
Specifications
| Chemical Name or Material | S-methyl 6a,9a-difluoro-11b-hydroxy-16a-methyl-3-oxo-17a-propionyloxy-androsta-1,4-diene-17beta-carbothioate |
| CAS | 73205-13-7 |
| Analyte or Component Names | S-methyl 6a,9a-difluoro-11b-hydroxy-16a-methyl-3-oxo-17a-propionyloxy-androsta-1,4-diene-17beta-carbothioate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H32 F2 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Fluticasone Propionate Imp. D (EP), Fluticasone Propionate USP Related Compound D, Fluticasone Propionate USP RC D, (6alpha,11beta,16alpha,17alpha)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)-androsta-1,4-diene-17-carbothioic Acid S-Methyl Ester, RS 35909-00-00-0, Ticabesone propionate, CCI 18771, USP Fluticasone Propionate Related Compound D, Ph Eur Fluticasone Propionate Impurity D, BP Fluticasone Propionate Impurity D, S-methyl fluticasone propionate CCI18771 USP Related Compound D, Compound A (fluticasone propionate related), unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), dithio (GR 269949X), Fluticasone Propionate Related Compound D, Fluticasone Propionate Impurity D |
| InChI Formula | InChI=1S/C25H32F2O5S/c1-6-20(30)32-25(21(31)33-5)13(2)9-15-16-11-18(26)17-10-14(28)7-8-22(17,3)24(16,27)19(29)12-23(15,25)4/h7-8,10,13,15-16,18-19,29H,6,9,11-12H2,1-5H3/t13-,15+,16+,18+,19+,22+,23+,24+,25+/m1/s1 |
| SMILES | CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SC |
| IUPAC Name | [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-17-methylsulfanylcarbonyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?