Learn More
6-(tert-Butylamino)-1,3,5-triazine-2,4(1H,3H)-dione, TRC
Product Code. 30137787
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137787 100 mg 100mg
30004296 10 mg 10mg
30006748 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137787 Supplier Toronto Research Chemicals Supplier No. TRCB185955100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 309923-18-0
Molecular Formula C7 H12 N4 O2
Molecular Weight (g/mol) 184.1958
Synonym Terbuthylazine Metabolite CGA 324007, 1,3,5-Triazine-2,4(1H,3H)-dione, 6-[(1,1-dimethylethyl)amino]-, 6-[(1,1-Dimethylethyl)amino]-1,3,5-triazine-2,4(1H,3H)-dione, LM5, Terbuthylazine Metabolite LM5
IUPAC Name 6-(tert-butylamino)-1,3,5-triazine-2,4-diol
SMILES CC(C)(C)Nc1nc(O)nc(O)n1
Specifications
Analyte or Component Names Terbuthylazine Metabolite CGA 324007
Shipping Condition Room Temperature
InChI Formula InChI=1S/C7H12N4O2/c1-7(2,3)11-4-8-5(12)10-6(13)9-4/h1-3H3,(H3,8,9,10,11,12,13)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 184.096
Chemical Name or Material Terbuthylazine Metabolite CGA 324007