missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylpyridine-2-carboxylic Acid, TRC

Product Code. 30411454
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30411454 25 g
30419349 5 g
30416212 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411454 Supplier Toronto Research Chemicals Supplier No. TRCP33660025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39774-28-2
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
Synonym 6-Phenylpicolinic Acid, 6-Phenyl-2-pyridinecarboxylic Acid
IUPAC Name 6-phenylpyridine-2-carboxylic acid
SMILES OC(=O)c1cccc(n1)c2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H9NO2/c14-12(15)11-8-4-7-10(13-11)9-5-2-1-3-6-9/h1-8H,(H,14,15)
Formula Weight 199.0633
Chemical Name or Material 6-Phenylpyridine-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.